Methyl 6-bromo-3-chloro-2-fluoro-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

432640-1G 432640-5G 432640-500MG
77638-746EA 591 USD
77638-746 77655-712 77650-728
Methyl 6-bromo-3-chloro-2-fluoro-α-oxobenzeneacetate ≥95%
Methyl 6-bromo-3-chloro-2-fluoro-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: C₉H₅BrClFO₃
Storage Temperature: Ambient
CAS Number: 2091687-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR