Methyl 3-chloro-2,4,6-trifluoro-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

432539-500MG 432539-5G 432539-1G
77652-804EA 462 USD
77652-804 77659-292 77642-764
Methyl 3-chloro-2,4,6-trifluoro-α-oxobenzeneacetate ≥95%
Methyl 3-chloro-2,4,6-trifluoro-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: C₉H₄ClF₃O₃
Storage Temperature: Ambient
CAS Number: 2090944-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR