2,2-Difluoro-1-(5-methyl-2-thienyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

432305-5G 432305-1G 432305-500MG
77629-274EA 1484.23 USD
77629-274 77638-744 77652-802
2,2-Difluoro-1-(5-methyl-2-thienyl)ethanone ≥97%
2,2-Difluoro-1-(5-methyl-2-thienyl)ethanone

Precaución: For Research Use Only

Formula: 176.19 g/mol
MW: C₇H₆F₂OS
Storage Temperature: Ambient
CAS Number: 1565033-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR