Ethyl α-oxo-5-phenyl-2-thiopheneacetate ≥95%

Supplier: AOBChem USA

Warning

432088-5G 432088-25G 432088-1G 432088-500MG
77629-272EA 1484.23 USD
77629-272 77635-800 77638-740 77646-784
Ethyl α-oxo-5-phenyl-2-thiopheneacetate ≥95%
Ethyl α-oxo-5-phenyl-2-thiopheneacetate

Precaución: For Research Use Only

Formula: 260.31 g/mol
MW: C₁₄H₁₂O₃S
Storage Temperature: Ambient
CAS Number: 100006-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR