Ethyl 7-methyl-α-oxobenzo[b]thiophene-2-acetate ≥95%

Supplier: AOBChem USA

Warning

432082-1G 432082-5G 432082-500MG
77638-738EA 522 USD
77638-738 77655-710 77646-782
Ethyl 7-methyl-α-oxobenzo[b]thiophene-2-acetate ≥95%
Ethyl 7-methyl-α-oxobenzo[b]thiophene-2-acetate

Precaución: For Research Use Only

Formula: 248.3 g/mol
MW: C₁₃H₁₂O₃S
Storage Temperature: Ambient
CAS Number: 1888747-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR