Ethyl 3-methyl-α-oxobenzo[b]thiophene-2-acetate ≥95%

Supplier: AOBChem USA

Warning

432053-5G 432053-1G 432053-500MG
77659-286EA 2417 USD
77659-286 77642-760 77646-780
Ethyl 3-methyl-α-oxobenzo[b]thiophene-2-acetate ≥95%
Ethyl 3-methyl-α-oxobenzo[b]thiophene-2-acetate

Precaución: For Research Use Only

Formula: 248.3 g/mol
MW: C₁₃H₁₂O₃S
Storage Temperature: Ambient
CAS Number: 100006-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR