Methyl α-oxobenzo[b]thiophene-2-acetate ≥95%

Supplier: AOBChem USA

Warning

432021-1G 432021-5G 432021-500MG
77638-736EA 633 USD
77638-736 77655-708 77650-724
Methyl α-oxobenzo[b]thiophene-2-acetate ≥95%
Methyl α-oxobenzo[b]thiophene-2-acetate

Precaución: For Research Use Only

Formula: 220.25 g/mol
MW: C₁₁H₈O₃S
Storage Temperature: Ambient
CAS Number: 214061-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR