Ethyl 5-bromo-4-methyl-α-oxo-2-thiopheneacetate ≥95%
Supplier: AOBChem USA
|
Warning
|
432018-5G
432018-1G
432018-500MG
432018-25G
77633-120EA
1171
USD
77633-120
77638-734
77652-800
77659-284
Ethyl 5-bromo-4-methyl-α-oxo-2-thiopheneacetate ≥95%
Ethyl 5-bromo-4-methyl-α-oxo-2-thiopheneacetate
Precaución: For Research Use Only
Formula:
277.14 g/mol MW: C₉H₉BrO₃S Storage Temperature: Ambient |
CAS Number:
1537376-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...