1-(8-Isoquinolinyl)-1-propanone ≥95%

Supplier: AOBChem USA

Warning

431675-5G 431675-1G 431675-500MG
77629-270EA 1484.23 USD
77629-270 77638-730 77646-778
1-(8-Isoquinolinyl)-1-propanone ≥95%
1-(8-Isoquinolinyl)-1-propanone

Precaución: For Research Use Only

Formula: C₁₂H₁₁NO
Storage Temperature: Ambient
CAS Number: 1053656-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR