[2-(1-Methylethyl)phenyl]phenylmethanone ≥95%

Supplier: AOBChem USA

Warning

431073-5G 431073-1G 431073-25G 431073-500MG
77633-114EA 1381 USD
77633-114 77638-728 77655-706 77646-776
[2-(1-Methylethyl)phenyl]phenylmethanone ≥95%
[2-(1-Methylethyl)phenyl]phenylmethanone

Precaución: For Research Use Only

Formula: 224.3 g/mol
MW: C₁₆H₁₆O
Storage Temperature: Ambient
CAS Number: 19103-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR