(5-Ethyl-2-methoxyphenyl)(phenyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

429748-5G 429748-25G 429748-1G 429748-500MG
77629-268EA 1484.23 USD
77629-268 77655-704 77642-756 77650-720
(5-Ethyl-2-methoxyphenyl)(phenyl)sulfane ≥97%
(5-Ethyl-2-methoxyphenyl)(phenyl)sulphane

Precaución: For Research Use Only

Formula: 244.3519 g/mol
MW: C₁₅H₁₆OS
Storage Temperature: Ambient
CAS Number: 847348-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR