Tert-butyl 4-(2,4,6-trifluorobenzoyl)piperidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

429708-5G 429708-1G 429708-500MG
77629-266EA 1484.23 USD
77629-266 77638-726 77646-774
Tert-butyl 4-(2,4,6-trifluorobenzoyl)piperidine-1-carboxylate ≥95%
Tert-butyl 4-(2,4,6-trifluorobenzoyl)piperidine-1-carboxylate

Precaución: For Research Use Only

Formula: 343.3408 g/mol
MW: C₁₇H₂₀F₃NO₃
Storage Temperature: Ambient
CAS Number: 2680772-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR