tert-Butyl 4-[hydroxy(phenyl)methyl]piperidine-1-carboxylate ≥95%
Supplier: AOBChem USA
|
Warning
|
428862-5G
428862-1G
428862-25G
428862-500MG
77633-112EA
1344
USD
77633-112
77638-722
77655-702
77646-772
tert-Butyl 4-[hydroxy(phenyl)methyl]piperidine-1-carboxylate ≥95%
tert-Butyl 4-[hydroxy(phenyl)methyl]piperidine-1-carboxylate
Precaución: For Research Use Only
Formula:
C₁₇H₂₅NO₃ MW: 291.3853 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12912653 CAS Number: 269740-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...