Rochester-Pean Hemostatic Forceps
Supplier: WORLD PRECISION INSTRUMENTS LLC
501325 501325-G 501706 501707 501708 501708-G
77662-596EA 46.22 USD
77662-596 77662-598 77662-676 77662-678 77662-680 77662-682
Rochester-Pean Hemostatic Forceps
Forceps
Rochester-Pean forceps may be used in a variety of surgical procedures for clamping blood vessels or other tissues. The solid grip offers surgeons a strong, secure grip, necessary when controlling bleeding.
  • Used for controlling bleeding and gripping tissue securely
  • 16 to 19 cm long
  • Locking handle
  • Stainless steel
The serrated, grooves along the jaws are designed for an effective grip on tissue to prevent tissue from slipping out of the forceps during use. These hemostatic forceps are available in a variety of sizes, have a locking handle and are constructed of stainless steel. Our premium line is constructed from German stainless steel.