PCR Plates without Skirt - Low Profile
Supplier: SARSTEDT INC
72.1977.202 72.1977.232
77588-594CS 247.33 USD
77588-594 77588-596
PCR Plates without Skirt - Low Profile
Microplates PCR Plates
SARSTEDT PCR consumables are compatible with standard thermocyclers. Uniform and extremely thin well walls facilitate consistently homogeneous and rapid heat transfer. During production, each individual well is tested to ensure it is 100% leak-proof.
  • Volume: 0.1 ml
  • 96-well, low profile design
  • Polypropylene material
  • Easy to cut if 24-well or 48-well formats are required
  • Optimized white color variant for maximum detection sensitivity in qPCR
  • For low-volume reactions and fast PCR
  • Constant well and plate uniformity minimizes data variability
SARSTEDT PCR plates are 'PCR Performance Tested' certified and guaranteed to be DNA-free, free from DNase/RNase, and free from PCR inhibitors.