Deep Well Plates, 96-Well
Supplier: SARSTEDT INC
82.1969.002 82.1970.002 82.1971.002 82.1972
77589-214CS 450.42 USD
77589-214 77589-216 77589-218 77589-220
Deep Well Plates, 96-Well
Microplates Deep Well Microplates
SARSTEDT offers deep well plates for a variety of applications.
  • Optimized shape for improved mixing properties and maximum sample recovery
  • Highly resistant to most solvents and mechanical stress
  • Fewer pipetting errors due to alphanumeric referencing
SARSTEDT plates are ideal for sample processing on automated systems. Round well plages are free from DNA/DNase/RNase, free from pyrogens/endotoxins. Square well plates are free from DNA/DNase/RNase/PCR inhibitors. The square well plates are compatible with most devices designed for automated sample isolation (such as the Thermo Scientific™ KingFisher™ Flex).