SPORTROL® Spore Suspensions
Supplier: Crosstex
NMSUS-06 NMSUN-06 NMSUP-06
19872-024EA 716.11 USD
19872-024 19872-022 19872-026
SPORTROL® Spore Suspensions
Sterilization Indicators
Spore suspensions are pure suspensions of viable spores from specific organisms of known resistance to various sterilization processes.
Use these biological indicators to provide validation that bacterial spores have been completely destroyed. Simply process the indicator with the load in your sterilizer. No bacterial growth after culturing indicates that your sterilization process is effective. Growth indicates ineffective sterilization. Spores are suspended in water for injection and provided in 10mL pharmaceutical vials. Available in a population of 106 spores per 0.1mL. Spore suspensions have a shelf life of 18 months when stored under labeled conditions.
Certificaciones: Meet USP and ISO 11138-1 standards.