Demeter Pipettes
Supplier: SARSTEDT INC
86.1686.025 86.1686.225
77589-588CS 231.9 USD
77589-588 77589-590
Demeter Pipettes
Pipettes Haemometer Pipettes
Dosing, transferring, and measuring liquids are routine procedures in almost every laboratory. SARSTEDT offers a wide range of products for every application, from high-quality pipette tips and serological pipettes to transfer pipettes, for reliable and reproducible results.
  • Working volume: 1.1 ml
  • Sterile polystyrene material
  • Optimized graduations for creating dilution series
  • For bacteriological testing in food laboratories
  • Cotton plugs for a lower risk of contamination
SARSTEDT Demeter pipettes provide controlled measuring of liquid.