PCR Strips - High Profile
Supplier: SARSTEDT INC
72.991.002 72.991.102 72.994.002 72.994.992
77589-014CS 652.45 USD
77589-014 77589-016 77589-020 77589-022
PCR Strips - High Profile
Tubes PCR Tubes
SARSTEDT PCR consumables are compatible with standard thermocyclers. Uniform and extremely thin well walls facilitate consistently homogeneous and rapid heat transfer. During production, each individual well is tested to ensure it is 100% leak-proof.
  • High profile design
  • Supplied with closures attached
  • Polypropylene material
  • Optimal heat transfer through thin-walled reaction chamber
  • Integrated contamination protection; can be opened without touching the inside surface of the lid
  • Color variants for optimal organization in the laboratory
SARSTEDT PCR tubes are 'PCR Performance Tested' certified and guaranteed to be DNA-free, free from DNase/RNase, and free from PCR inhibitors.
Assorted color option includes a mix of Red, Green, Blue, Purple, and Yellow.