PCR Plates with Half Skirt - Low Profile
Supplier: SARSTEDT INC
72.1981 72.1981.010 72.1982.202
77588-624CS 483.41 USD
77588-624 77588-626 77588-628
PCR Plates with Half Skirt - Low Profile
Microplates PCR Plates
SARSTEDT PCR consumables are compatible with standard thermocyclers. Uniform and extremely thin well walls facilitate consistently homogeneous and rapid heat transfer. During production, each individual well is tested to ensure it is 100% leak-proof.
  • Volume: 0.1 ml
  • 96-well, low profile design
  • Polypropylene material
  • Optimized white color variant for maximum detection sensitivity in qPCR
  • For low-volume reactions and fast PCR
  • PCR plate 72. 1982.252 compatible with Roche LightCycler® 480
  • Constant well and plate uniformity minimizes data variability
SARSTEDT PCR plates are 'PCR Performance Tested' certified and guaranteed to be DNA-free, free from DNase/RNase, and free from PCR inhibitors.