Caps for Reaction Tubes
Supplier: SARSTEDT INC
65.697 65.109 65.723
77588-148CS 367.71 USD
77588-148 77588-030 77588-228
Caps for Reaction Tubes
Closures Test/Sample Tube Closures
High-quality, safe and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • LDPE closure material
  • Flat stopper for space-saving storage
  • High stopper for easy opening and closing
Particularly suitable for automated closing, attached caps ensure reliable assignment to the container.