SafeSeal Reaction Tubes, 5 ml
Supplier: SARSTEDT INC
72.701.400 72.701.500
77588-874CS 371.36 USD
77588-874 77588-876
SafeSeal Reaction Tubes, 5 ml
Tubes PCR Tubes
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • SafeSeal polypropylene caps attached
  • Polypropylene tube material
  • Easy handling of filling quantities between 2 and 5 ml
  • Reliable SafeSeal closure
  • 3 purity grades to meet all requirements
  • High centrifugation up to 20,000×g
This broad product portfolio in the field of test and micro tubes includes options with round, conical or flat base; with or without skirted base; with push cap or screw cap; with or without print or labels.