Halsey Needle Holders
Supplier: WORLD PRECISION INSTRUMENTS LLC
14110 14110-G
77661-728EA 63.15 USD
77661-728 77661-730
Halsey Needle Holders
Forceps
Halsey needle holders are used to hold and manipulate surgical needles during procedures. Halsey needle holders have a scissor-like jaw design with a flat, blunt tip.
They have a locking mechanism to secure the jaws as they grasp the needle firmly. While Halsey needle holders are available in sizes from 4 to 7", these are 5.5" long.