SHOWA CN741 Chemical Resistance Gloves
Supplier: Showa
CN741L-09 CN741M-08 CN741S-07 CN741XL-10 CN741XXL-11
77623-506BG 55.75 USD
77623-506 77623-508 77623-510 77623-512 77623-514
SHOWA CN741 Chemical Resistance Gloves
Gloves
The SHOWA CN741 features a high level of chemical resistance. At 380 mm in length, the glove is designed with a grip wave pattern to ensure a secure grip during use, even in challenging conditions. The interior of the glove incorporates innovative technology. The flocking particles help to distribute moisture, making it easier to put on and take off the gloves even after extended wear. The SHOWA CN741 also features Eco Best Technology, making it a biodegradable glove.
  • Chemical-resistant
  • Biodegradable and nitrile coating
  • Forearm protection
  • Oil-resistant
  • Impermeable
  • Easy donning and doffing
  • Durable
Features:
Eco Best Technology® (EBT)
Unlined
Wave pattern finish
Industries: Food
Trades and applications:
Chemical spray and treatment
Handling light chemicals in agriculture, horticulture
Light assembly of oil-coated pieces
Coating preparation
Washing and cleaning
Certificaciones: Cat III, EN 388:2016+A1:2018, EN ISO 374-1:2016+A1:2018/Type, EN ISO 374-5:2016, Food Contact, EN 420:2003+A1:2009.
Envase: 12 pairs per polybag, 6 polybags per case and 72 pair per case.