SHOWA 729 Gloves, Nitrile Coating
Supplier: Showa
729-07 729-08 729-09 729-10 729-11
77623-476CS 189.83 USD
77623-476 77623-478 77623-480 77623-482 77623-484
SHOWA 729 Gloves, Nitrile Coating
Gloves
SHOWA 729 offers a diamond grip surface texture gives the wearer enhanced grip, to make wet work safer and easier. The ergonomic design maximizes comfort to aid precision. The 100% nitrile glove provides protection against a broad range of oils, solvents and light chemicals.
  • Chemical-resistant
  • Water-resistant
  • Forearm protection
  • Oil-resistant
  • Flexible
Used in automotive, chemical and mining industries.
Trades and applications:
Chemical spray and treatment
Light chemical handling in horticulture, agriculture
Light assembly of oil-coated pieces
Painting and spray workshops
Coating preparation
Washing and cleaning
Envase: 12 pair per polybag, 12 polybags per case, 144 pair per case.