Webster Needle Holders
Supplier: WORLD PRECISION INSTRUMENTS LLC
14109 14109-G 501843 501929
77661-724EA 63.15 USD
77661-724 77661-726 77662-878 77662-954
Webster Needle Holders
Forceps
Webster needle holders are surgical instrument use to hold and manipulate suturing needles. While Webster needle holders are often associated with microsurgery and microvascular procedures, they can be used in a variety of surgical contexts where precise needle handling is required.
  • Extra fine or rine straight tips
  • Serrated or smooth jaws
  • 1.2 mm tips
  • Lock
  • Surgical grade stainless steel or premium German steel
Webster needle holders have fine tips for precise control when grasping needles. The fine tips make it easier to handle and manipulate small and fine needles used in microsurgery and microvascular procedures. We also offer extra fine tips for smaller needles. Like most needle holders, Webster needle holders have a locking handle for a secure hold on the needle, so that it remains stable during suturing, reducing the risk of slipping or accidental needle sticks.
The jaws of some WPI Webster needle holders are lightly textured for a solid grip. Others have smooth jaws to minimize tissue damage. The ratchet handle lets you adjust the tension on the needle, ensuring a secure grip while suturing.
These surgical instruments are manufactured from surgical grade stainless steel or German grade stainless steel, both of which are suitable for sterilization by autoclaving or chemical sterilant. To ensure a long life for your surgical instruments, always clean them promptly.