Chemical Resistant Gloves, CS711
Supplier: Showa
CS711L-09 CS711M-08 CS711S-07 CS711XL-10 CS711XXL-11
76521-156BG 64.79 USD
76521-156 76521-154 76521-152 76521-158 76521-160
Chemical Resistant Gloves, CS711
Gloves
The CS711 chemical-resistant gloves offer enhanced grip, comfort, and safety. Forearm protection and nitrile material keep oil and dust from entering the flove. A seamless liner and natural latex-free material ensures comfort and reduces irritation. Even in oily or wet wroking enviroments, the foamed nitrile palm coating provides the user with tactile precision and protection.
  • Chemical and oil resistant
  • Extra grip
  • Seamless knit
  • Robust durability
  • Forearm protection
  • Double coated nitrile
  • Polyester liner
  • Foam grip
  • Microporous nitrile coating
The glove is suitable for commercial fishing, oil and gas, aerospace, chemical, construction, printing, automotive.
Chemical spray and treatment, light chemical handling in horticulture, agriculture, coating preparation, laboratory, pharma and analysis, painting and spray workshops, fruit and vegetable processing, grain, mill and starch products.
Certificaciones: CE, EN 388:2016+A1:2018, EN ISO 374-1:2016+A1:2018/Type A, EN ISO 374-5:2016, EN 407:2004 certified.
Envase: Ten pair per polybag, six polybags per case, 60 pair per case.