Cole-Parmer® SHM-200 Series Stirring/Heating Mantles, Antylia Scientific
Supplier: Antyila Scientific
04641-20 04641-50 04641-00 04641-30 04641-10 04641-40
33750-356EA 1621.01 USD
33750-356 33750-447 33750-298 33750-389 33750-323 33750-414
Cole-Parmer® SHM-200 Series Stirring/Heating Mantles, Antylia Scientific
Heaters Heating Mantles
Provides powerful stirring speeds up to 520 rpm and heating up to 450 °C.
  • Bidirectional stirring with stir bar auto capture function
  • Thermal insulating cartridge provides maximum heat transfer
  • Chemical resistant polypropylene housing is 'safe to the touch'
The added functionality of stirring widens the range of applications. Stirring operation is simply achieved by placing the correct size stir bar into the flask, turning the stir control to the minimum speed, and ramping up slowly. If the magnetic coupling to the stir bar is lost for any reason, theMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable