Ammonium hexafluorotitanate ≥99.99% (trace metals basis), SPEX Infinitas™

Supplier: SPEX CERTIPREP LLC
Danger

IFC-TI-4A-10 IFC-TI-4A-25
SXIFC-TI-4A-10EA 75 USD
SXIFC-TI-4A-10 SXIFC-TI-4A-25
Ammonium hexafluorotitanate ≥99.99% (trace metals basis), SPEX Infinitas™
Ammonium hexafluorotitanate
Formula: H₈F₆N₂Ti
MDL Number: MFCD00010888
CAS Number: 16962-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR