Lead(II) nitrate ≥99.99% (trace metals basis)
Supplier: SPEX CERTIPREP LLC
|
Danger
|
Synonyms:
Lead dinitrate
IFC-PB-1A-25
IFC-PB-1A-50
SXIFC-PB-1A-25EA
130
USD
SXIFC-PB-1A-25
SXIFC-PB-1A-50
Lead(II) nitrate ≥99.99% (trace metals basis)
Lead(II) nitrate
Formula:
Pb(NO₃)₂ MW: 331.21 g/mol Melting Pt: 470 °C (878 °F) |
MDL Number:
MFCD00011153 CAS Number: 10099-74-8 UN: 1469 ADR: 5.1 II Merck Index: 13,05432 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...