Chromium(III) hydroxide ≥99.99% (trace metals basis)

Supplier: SPEX CERTIPREP LLC

Synonyms: Chromium trihydroxide

IFC-CR-2A-10 IFC-CR-2A-50
SXIFC-CR-2A-10EA 105 USD
SXIFC-CR-2A-10 SXIFC-CR-2A-50
Chromium(III) hydroxide ≥99.99% (trace metals basis)
Chromium(III) hydroxide
Formula: H₃CrO₃
MDL Number: MFCD00049432
CAS Number: 1308-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR