VWR® 700 Portable Conductivity Meters
Supplier: VWR International
77618-970 77618-964 77618-966 77618-968
77618-970EA 903.93 USD
77618-970 77618-964 77618-966 77618-968
VWR® 700 Portable Conductivity Meters
Electrochemistry Meters Conductivity Meters
The VWR-EC700 meter provide accurate and reliable measurements of conductivity, resistivity, TDS, salinity and temperature in any environment.
  • High resolution LCD display (3.5")
  • Three point calibration with Standard Recognition
  • IP65 waterproof
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Ability to save 500 data sets for each parameter (GLP-compliant)
  • Rechargeable battery
  • Settable parameters, including cell constant, temperature compensation coefficient, and TDS factor
The easy to use VWR-EC700 portable meters provide accurate conductivity measurements inside and outside the lab. The large, backlit LCD display allows results to easily be viewed during operation. The carrying case allows for easy transport of the meter and accessories.
Información para pedidos: Three-year meter warranty. For application specific recommendations, please reference 'VWR® electrode selection guide' located in the related information section.
Información de suministro: All meters include carrying case, two electrode holders, meter silicone rubber case, and hand strap.