Glas-Col® Heating Tapes
Supplier: Glas-Col
103AFET0.67 103ASET14 103AFET0.65 103ASET13 103ADET0510 103AFET0.610 103AFET0.66 103ADET110 103AFET0.68 103AFET0.69 103ADET02510
80078-788EA 376.33 USD
80078-788 80078-798 80078-784 80078-796 80078-780 80078-794 33730-284 80078-782 80078-790 80078-792 80078-778
Glas-Col® Heating Tapes
Tapes
Heat odd lengths of glass tubes and metal pipes for both permanent and temporary applications with these heating tapes.
The tapes are ideal for use in high-moisture areas, are resistant to most laboratory environmental conditions, and will easily pass a 1000 V test for electrical insulation. Ribbon-type heating elements running lengthwise in the tape are encased in a durable glass braid and are impregnated and covered with tough silicone rubber. The glass braid and silicone rubber serve to insulate the stranded terminal leads at one end and waterproof the fittings at both ends. The attached cord has a two-blade plug and a single end termination. Temperature limit: 249 °C (480 °F).
Precaución: Note: Power ratings are for tapes not covered by insulation when installed.