Cole-Parmer® HM-200 Series Heating Mantles, Antylia Scientific
Supplier: Antyila Scientific
36002-04 36002-00 36002-08 36002-16 36002-12 36002-20 36002-24 36002-28
33750-039EA 744.68 USD
33750-039 33750-017 75994-944 75994-946 76045-610 76045-612 76045-614 76045-864
Cole-Parmer® HM-200 Series Heating Mantles, Antylia Scientific
Heaters Heating Mantles
Thermal insulating cartridge provides maximum heat transfer.
  • Chemical resistant polypropylene housing is ‘safe to the touch’
Quickly and safely heat round-bottom flasks to 450 °C (842 °F). These simple-to-use heating mantles feature a built-in power controller with dial markings from 1 to 10 for repeatability. An amber-colored ‘heater on’ light and a clear-colored ‘power on’ light indicate when the heating mantle is in use. The replaceable coiled heating element suspended within a thermal insulating cartridge provides maximum heat transfer and support.
Unique ventilation slots and ceramic fiber insulation keep the chemical-resistant polypropylene exterior cool even while in use. A safety earth (ground) screen and double fuses provide added safety.Supplied with power cord with US plug.
Información para pedidos: Units are supplied with brackets for 1.3 cm diameter support rods (order does not include clamps or support rods). Flask not included. Model 33750-094 has 2 circuits for upper and lower level heating.