VWR® Nano Spectrophotometer LS
Supplier: VWR International
77610-710 77610-712
77610-710EA 7143.55 USD
77610-710 77610-712
VWR® Nano Spectrophotometer LS
Spectrophotometers and Photometers Biophotometers
The VWR® Nano Spectrophotometer LS is the affordable choice for laboratories needing to determine nucleic acid and protein concentrations and purity of samples.
  • Ideal for basic DNA/RNA/protein quantification and purity affordable LED and silicon detector technology
  • Fixed 0.5 mm path length, detect up to 2500 ng/µl of dsDNA
  • Silicon photocell window material
The LED light source and silicon photocell sensor provide readings at 260 and 280 nm. Samples from 1.0 to 2.0 µl are loaded directly on the pedestal. Results are provided in about six seconds. The software automatically calculates and displays 260/280 ratios. Data can be printed using the integrated thermal printer, saved in the instrument's onboard memory or exported via USB as an Excel file. A cuvette port allows for OD 600 readings and two optical glass cuvettes are included.
Detection time: <6 sec
Absorbance precision = 0.005 A