SIS-212 Softwall Portable Cleanrooms, Simplex
Supplier: Simplex Strip Doors
SIS0808E SIS0808P SIS1010S SIS0808S SIS1010E SIS1010P
89042-132EA 16764.27 USD
89042-132 89042-134 89042-136 89042-130 89042-138 89042-140
SIS-212 Softwall Portable Cleanrooms, Simplex
Prefabricated Rooms
System meets requirements for ISO Class 8 to ISO Class 5 (FED-STD-209E Class 100,000/M6.5 to Class 100/M3.5)
Softwalls are available in three materials: STD-Clear, ESD-Clear, and polysim. The STD-Clear material is a fire retardant, standard grade vinyl. The ESD-Clear material is a static dissipative vinyl with surface resistivity of 1.8x1010ohm/cm. STD- and ESD-Clear strips are .060mil thick and 20.3cm (8") wide. The polysim strips, made of a static dissipative non-outgassing polyurethane, are .040mil thick and 30.5cm (12”) wide.
Designed for fast construction and can be easily customized. Frames can be quickly assembled in modular sections then connected to each other with internal screws. Modules can be easily disassembled or expanded. Enclosures can be fitted with HEPA filters and modular air conditioning units, or can be quickly adapted to existing ventilating and dust collection systems. The frame system is constructed of heavy-duty 5.4cm (21/8") thick anodized aluminum with knock-in corners for easy assembly. Available with fully integrated ceiling modules that have a 5.1cm (2") T-grid and neoprene closed-cell foam gaskets for an airtight seal. Softwalls consist of three sides with .040mil clear panels and one side with alternating clear/frosted strips for easy entrance and exiting.
Certificaciones: Stock sizes listed below meet ISO Class 7 (FED-STD-209E Class 10,000/M5.5) standards.
Información para pedidos: System includes ceiling tiles; Class 100 T-8 energy efficient, 100% sealed cleanroom lights; and fan-powered HEPA filter units rated ISO Class 5 (FED-STD-209E Class 100/M3.5) that are 99.99% efficient on particles of 0.3µm or larger. Standard height is 2.4m (8'). For custom sizes, different classes of rooms, or for other curtain options, contact your Avantor sales representative for more information.