Biosafety Cabinets, Cell Logic Class II, Type B2
Supplier: Labconco
323481002 323481012 323481102 323481112 323681002 323681012 323681102 323681112 343481002 343481012 343481102 343481112 343681002 343681012 333481002 333481012 333481102 333481112 333681002 333681012 333681102 333681112 343681102 343681112
77574-190EA 25125 USD
77574-190 77574-398 77574-192 77574-400 77574-294 77574-502 77574-296 77574-504 77574-198 77574-406 77574-200 77574-408 77574-302 77574-510 77574-194 77574-402 77574-196 77574-404 77574-298 77574-506 77574-300 77574-508 77574-304 77574-512
Biosafety Cabinets, Cell Logic Class II, Type B2
Fume Hoods and Enclosures Biological Safety Cabinets
Cell logic Class II, Type B2 Biosafety Cabinets (BSCs) offer comprehensive personnel, product and environmental protection from hazardous particulates, including risk group agents requiring BSL 1-4 containment. Type B2 BSCs, sometimes referred to as 100% exhaust or total exhaust BSCs require a dedicated exhaust system and remote blower for each cabinet. Cell Logics are designed to meet the specific needs of cell research. Models feature a modified sash to accommodate a microscope and/or a temperature-controlled work area on the work surface to maintain cell viability. All models ensure the highest standard of safety and performance.
  • Safe Class II airflows (100% exhaust, remote blower required) that provide personnel, product, and environmental protection. Cabinet interior provides ISO Class 5 (Grade A) or better working conditions
  • Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe downflow - even as HEPA filters load. Optimized airflow minimizes energy use and noise output to industry-leading levels
  • Logic OS with 5” line-of-sight touchscreen display. Access cabinet information such as filter life, alerts and alarms, data logs. Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet
  • Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work
  • Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work
  • Remote Communication Board for direct communication with remote blower and/or building management system featuring dry contact relays, USB data output and RS-232 RJ45 data output
  • Full, six year warranty on all Class II biosafety cabinets
Logic BSCs maximize safety, performance, and comfort for users. Constant Airflow Profile™ (CAP) technology and BalanceFlow™ plenum technology ensure optimal airflow control. Every Logic is powered by Logic OS - an advanced software platform with touchscreen interface located right at eye-level within the cabinet. All B2 models feature a remote communication board for remote blower communication and inflow/downflow airflow monitors.
Optional configuration include Scope-Ready™ Package consisting of 7.0W× 10.5" H (18×27 cm) sash cutout and Pure-Vu™ Seal with powder-coated steel collar. Stand-Still™ Isolation Platform is required and sold separately. Alternatively the Temp-Zone™ Package consisting of temperature-controlled work area, 10.5W×12.0" D (27×30 cm) located on the left side of the work surface. Water circulator is required (not included). Another optional configuration is the Telescoping base stand with shelf and leveling feet.
Accessory package include UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals. The Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach.
Certificaciones: ISO Class 5 (Grade A) per ISO 14644-1; UL Standard 61010-1; CAN/CSA 22.2 No. 61010-1; NSF/ANSI 49 compliant.