PuriCare Procedure Stations
Supplier: Labconco
312320002 312320012 312321002 312321012 312420002 312420012 312421002 312421012 312520002 312520012 312521002 312521012 312620002 312620012 312621002 312621012
77574-172EA 13230 USD
77574-172 77574-380 77574-174 77574-382 77574-234 77574-442 77574-236 77574-444 77574-276 77574-484 77574-278 77574-486 77574-338 77574-546 77574-340 77574-548
PuriCare Procedure Stations
Fume Hoods and Enclosures Biological Safety Cabinets
PuriCare procedure stations offer comprehensive personnel, product and environmental protection from hazardous particulates, including risk group agents requiring BSL 1-4 containment. These Class II, Type A2 biosafety cabinets are ideal for small animal research activities including surgical procedures, cage changing, weighing, and anesthesia delivery. All models ensure the highest standard of safety and performance and are listed to NSF/ANSI 49.
  • Safe Class II airflows (approx. 70% recirculating, 30% exhausted) that provide personnel, product, and environmental protection
  • Cabinet interior provides ISO Class 5 (Grade A) or better working conditions
  • Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe cabinet airflows – even as HEPA filters load
  • Optimized airflow minimizes energy use and noise output to industry-leading levels
  • Removable, washable prefilters to protect HEPA longevity
  • Logic OS with 5" line-of-sight touchscreen display. Access cabinet information such as filter life, alerts and alarms, data logs
  • Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet
  • Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work
  • Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work
  • Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach
  • Full, six-year warranty on all Class II biosafety cabinets
Designed for application versatility, Logic A2 BSCs either exhaust HEPA-filtered air into the laboratory or can be connected to a laboratory exhaust system with applications involving chemical vapors or odors.
Logic BSCs maximize safety, performance, and comfort for users. At the core is Constant Airflow Profile™ (CAP) technology, which keeps users and their samples protected at all times. Now paired with BalanceFlow™ plenum technology, logic cabinets use up to 30% less energy and are up to 5 decibels quieter. Every Logic is powered by Logic OS - an advanced software platform with touchscreen interface located right at eye-level within the cabinet.
Optional configurations: The accessory package includes UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ cord and cable portals.
Certificaciones: NSF/ANSI Standard 49 listed; ISO Class 5 (Grade A) per ISO 14644-1; UL Standard 61010-1; CAN/CSA 22.2 No. 61010-1.