LED Sign Projectors, 60 W
Supplier: Brady Worldwide
177626 177627 177628 177629 177630
77603-578EA 1102.61 USD
77603-578 77603-580 77603-582 77603-584 77603-586
LED Sign Projectors, 60 W
Signs
Mark the unmarkable with LED sign projectors from Brady. Ensure safety and showcase professionalism anywhere with projectors ideal for challenging and changing environments.
  • Compatible with Gobo E size slides
  • 60 W LED lamp
  • Two-year warranty
Mark the unmarkable with LED sign projectors from Brady. Ensure safety and showcase professionalism anywhere with projectors ideal for challenging and changing environments. Project a wide variety of signage onto surfaces where paint and tape can’t, including those that are hot, humid, dusty, grimy, uneven and irregular. Eliminate the prep, install, waste and special training needed with disposable tapes and paints. Maintenance-free operation thanks to no fans, motors or moving parts. Designed for rugged industrial environments and is waterproof, making it safe for washdowns. Ideal for projecting medium floor signage up to 6' wide at a distance of up to 40'. Accepts E size sign lenses. UL/cUL certified, IP 65 rated. 35000 hour bulb life.
Información de suministro: Also includes mounting yoke, power cord, and test Gobo.