HyFlex® 11-561 Cut-Resistant Gloves
Supplier: Ansell Healthcare
11561050 11561060 11561070 11561080 11561090 11561100 11561110 11561120
76478-862CS 1450.22 USD
76478-862 76478-864 76478-866 76478-868 76478-870 76478-872 76478-874 76478-876
HyFlex® 11-561 Cut-Resistant Gloves
Gloves
Ansell's lightest EN ISO Cut C and ANSI/ISEA 105-2024 Cut A3 glove (versus standard EN ISO Cut C and ANSI/ISEA 105-2024 Cut A3-rated gloves) offering 2 times greater cut resistance versus similar gloves made of standard HPPE yarn.
  • Ultra-lightweight design; 20% lighter for unparalleled comfort and dexterity
  • EN ISO Cut C and ANSI/ ISEA 105-2024 Cut A3 protection; 2x greater cut resistance
  • Improved FORTIX™ Technology offering up to 20% greater durability
The ultra-lightweight design of the HyFlex® 11-561 makes this EN ISO Cut C and ANSI/ISEA 105-2024 A3-rated safety glove 20% lighter. The gloves provide heightened defense, with 2 times greater cut resistance using Next Generation single HPPE yarn developed with INTERCEPT™ Cut Resistance Technology for reliable hand protection. Offering increased durability for extended use, improved FORTIX™ Technology delivers up to 20% more abrasion resistance for longer lasting handling in abrasive conditions.
HyFlex 11-561’s palm-dipped coating improves grip, while a reinforced thumb crotch defends against rapid wear. Certified harm-free, Dermatest® and Oeko-Tex® certification ensures these PPE gloves are skin-friendly and free of harmful substances. Touchscreen compatibility allows the wearer to work with multiple touchscreen devices while keeping gloves on.
Certificaciones: ANSI/ISEA 105-2024 Cut Level A3, ANSI/ISEA 105-2024 ABR Level 4, ANSI/ISEA 105-2024 PUN Level 4, Dermatologically Tested, Ecovadis label, EN 388:2016, EN ISO 21420:2020, Food Contact, OEKO-TEX® Centexbel, Category II.
Envase: Gloves are packaged 12 pairs per bag, 144 pairs per carton.