3,6-Dihydro-2H-pyran-4-yl trifluoromethanesulfonate 95%

Supplier: Ambeed
Danger

A1145138-1G A1145138-250MG
77619-980EA 58.71 USD
77619-980 77619-982
3,6-Dihydro-2H-pyran-4-yl trifluoromethanesulfonate 95%
3,6-Dihydro-2H-pyran-4-yl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₆H₇F₃O₄S
MW: 232.18 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09997709
CAS Number: 188975-30-6
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR