1-Naphthylamine ≥98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Aminonaphthalene

104120250 104121000
TS104120250EA 39 USD
TS104120250 TS104121000
1-Naphthylamine ≥98%
1-Naphthylamine
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 301°C (760mmHg)
Flash Pt: 157°C
Storage Temperature: Refrigerator
MDL Number: MFCD00004016
CAS Number: 134-32-7
EINECS: 205-138-7
UN: 2077
ADR: 6.1,III

Order Now

Specification Test Results

Appearance light yellow to light brown to dark purple crystalline powder
Infrared spectrum Conforms
GC ≥97.5 %
Melting point 48°C to 52°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR