B-[4-(2,2,3,3-Tetrafluoropropoxy)phenyl]boronic acid ≥97%

Supplier: AOBChem USA

Warning

427119-5G 427119-25G 427119-1G 427119-500MG
77629-256EA 1484.23 USD
77629-256 77635-798 77642-750 77646-768
B-[4-(2,2,3,3-Tetrafluoropropoxy)phenyl]boronic acid ≥97%
B-[4-(2,2,3,3-Tetrafluoropropoxy)phenyl]boronic acid

Precaución: For Research Use Only

Formula: 251.97 g/mol
MW: C₉H₉BF₄O₃
Storage Temperature: Ambient
CAS Number: 958453-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR