4-Cyclopropyl-2-fluorobenzonitrile ≥95%

Supplier: AOBChem USA

Warning

426814-5G 426814-1G 426814-500MG
77629-254EA 1484.23 USD
77629-254 77642-748 77650-714
4-Cyclopropyl-2-fluorobenzonitrile ≥95%
4-Cyclopropyl-2-fluorobenzonitrile

Precaución: For Research Use Only

Formula: C₁₀H₈FN
Storage Temperature: Ambient
CAS Number: 1394068-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR