3,4-Dibromo-2-iodo-6-methylaniline ≥97%

Supplier: AOBChem USA

Warning

426784-5G 426784-1G 426784-25G 426784-500MG
77629-252EA 1642.85 USD
77629-252 77638-718 77655-698 77646-766
3,4-Dibromo-2-iodo-6-methylaniline ≥97%
3,4-Dibromo-2-iodo-6-methylaniline

Precaución: For Research Use Only

Formula: C₇H₆Br₂IN
Storage Temperature: Ambient
CAS Number: 3031808-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR