(3-Iodo-5-methylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

426691-5G 426691-1G 426691-500MG 426691-25G
77633-110EA 1441 USD
77633-110 77638-716 77652-794 77655-696
(3-Iodo-5-methylphenyl)trimethylsilane ≥95%
(3-Iodo-5-methylphenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 290.2160 g/mol
MW: C₁₀H₁₅ISi
Storage Temperature: Ambient
CAS Number: 3031788-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR