1-(5-Methylthiophen-3-yl)cyclopentanol ≥97%

Supplier: AOBChem USA

Warning

426050-5G 426050-1G 426050-500MG
77629-250EA 1484.23 USD
77629-250 77638-714 77646-764
1-(5-Methylthiophen-3-yl)cyclopentanol ≥97%
1-(5-Methylthiophen-3-yl)cyclopentanol

Precaución: For Research Use Only

Formula: 182.283 g/mol
MW: C₁₀H₁₄OS
Storage Temperature: Ambient
CAS Number: 1486723-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR