Cyclohexyl(5-methylthiophen-3-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

426048-5G 426048-1G 426048-500MG
77629-248EA 1553.24 USD
77629-248 77638-712 77650-712
Cyclohexyl(5-methylthiophen-3-yl)methanol ≥95%
Cyclohexyl(5-methylthiophen-3-yl)methanol

Precaución: For Research Use Only

Formula: C₁₂H₁₈OS
Storage Temperature: Ambient
CAS Number: 1468485-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR