(5-Methylthiophen-3-yl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

426038-5G 426038-25G 426038-1G 426038-500MG
77633-108EA 1366 USD
77633-108 77655-694 77642-746 77650-710
(5-Methylthiophen-3-yl)(phenyl)methanone ≥95%
(5-Methylthiophen-3-yl)(phenyl)methanone

Precaución: For Research Use Only

Formula: 202.272 g/mol
MW: C₁₂H₁₀OS
Storage Temperature: Ambient
CAS Number: 1026012-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR