Cyclohexyl(5-methylthiophen-3-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

426034-5G 426034-1G 426034-500MG
77629-246EA 1484.23 USD
77629-246 77642-744 77646-762
Cyclohexyl(5-methylthiophen-3-yl)methanone ≥95%
Cyclohexyl(5-methylthiophen-3-yl)methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₆OS
Storage Temperature: Ambient
CAS Number: 1491209-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR